Skip to Content
Merck
All Photos(1)

Key Documents

6.18404

Supelco

Bromophenol blue

pH indicator solution 0.04%

Synonym(s):

Bromophenol Blue solution, 3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
MDL number:
UNSPSC Code:
41116100

Product Name

Bromophenol blue, 0.04% (indicator)

concentration

0.04% (indicator)

SMILES string

[Na+].Oc1c(Br)cc(cc1Br)C(=C2/C=C(Br)C(=O)C(Br)=C2)\c3ccccc3S([O-])(=O)=O

InChI

1S/C19H10Br4O5S.Na/c20-12-5-9(6-13(21)18(12)24)17(10-7-14(22)19(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28;/h1-8,24H,(H,26,27,28);/q;+1/p-1

InChI key

TWKDJMRHSLWRFI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service