Skip to Content
Merck
All Photos(6)

Key Documents

CWA-1092

Sigma-Aldrich

Anti-Human Macrosialin/CD68 (Y1/82A) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, (Sold Separately))

Synonym(s):

Macrosialin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Y1/82A, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 37.41 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD68(968)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone Y1/82A is a mouse monoclonal antibody that detects Macrosialin (CD68).

Immunogen

PHA-stimulated Human peripheral blood mononuclear cells (PBMC).

Application

Quality Control Testing

Evaluated by Flow Cytometry in Human monocytes, intracellularly stained.

Flow Cytometry Analysis:Intracellular staining of one million Human monocytes was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1092, Anti-Human Macrosialin / CD68 / LAMP4 (Y1/82A) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Mouse IgG2b isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Macrosialin (UniProt: P34810; also known as Gp110, CD68) is encoded by the CD68 gene (Gene ID: 968) in human. CD68 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 22-319), a transmembrane domain (aa 320-345), and a short cytoplasmic tail (aa 345-354). It also contains a mucin-like domain (aa 23-140) and has two 30 amino acid long repeats (aa 70-99 and 100-129), proline-rich hinge (aa 140-152), and a LAMP-like domain. CD68 is highly expressed in blood monocytes and tissue macrophages. Its expression is also observed in lymphocytes, fibroblasts, and endothelial cells. It is also found in many tumor cells and allows them to attach to selectin on vascular endothelium, facilitating their dissemination to secondary sites. CD68 is mainly located in the endosomal/lysosomal compartment but can rapidly shuttle to the cell surface. In macrophages, it plays a role in their phagocytic activities, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. It binds to tissue- and organ-specific lectins or selectins, which allows for homing of macrophages to particular sites. It has a good predictive value as a prognostic marker of survival in cancer subjects. CD68 expression is shown to be upregulated in macrophages in response to inflammatory stimuli, such as exposure to oxLDL, lipopolysaccharide, or interferon-g. Hence, it can serve as a good histochemical/cytochemical marker of inflammation associated with the involvement of monocytes/macrophages. Its expression is not observed in dendritic cells; hence it can be used to immunohistochemically distinguish macrophages from dendritic cells.

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service