Skip to Content
Merck
All Photos(1)

Key Documents

700131P

Avanti

24S,27-dihydroxycholesterol

Avanti Research - A Croda Brand 700131P, powder

Synonym(s):

(25S)-cholest-5-en-3β,24S,27-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O3
CAS Number:
Molecular Weight:
418.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700131P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700131P

shipped in

dry ice

storage temp.

−20°C

General description

24S,27-dihydroxycholesterol differs from 24S-hydroxycholesterol in steroid nucleus by having an additional hydroxyl group. 24S,27-dihydroxycholesterol is derived from 24S-hydroxycholesterol in the presence of enzyme cholesterol 24-hydroxylase (P450 46A1).

Packaging

5 mL Amber Glass Screw Cap Vial (700131P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Mast et al.
Biochemistry, 42(48), 14284-14292 (2003-12-03)
The known activity of cytochrome P450 46A1 (P450 46A1) is 24(S)-hydroxylation of cholesterol. This reaction produces biologically active oxysterol, 24(S)-hydroxycholesterol, and is also the first step in enzymatic degradation of cholesterol in the brain. We report here that P450 46A1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service