MT51344295
Mettler-Toledo electrode cable
connector AK9, BNC-RCA
All Photos(1)
About This Item
UNSPSC Code:
41115600
NACRES:
NB.61
Recommended Products
manufacturer/tradename
Mettler-Toledo 51344295
General description
Mettler Toledo electrodes can easily be connected to various third-party instruments. All you have to do is select the appropriate cable. We offer a wide choice of cable lengths and plug combinations. For Mettler Toledo reference electrodes and temperature probes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)