주요 문서
27150-U
Supelco
Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100
volume 7 mL, clear glass vial, O.D. × H 17 mm × 60 mm, thread for 15-425
동의어(들):
PK100 7ML CLEAR VIAL,SCREW TOP,SOLID CAP W/PTFE LINER
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122001
NACRES:
NB.11
material
F217/PTFE cap liner
PTFE liner
clear glass vial
green melamine resin cap (solid-top)
특징
closure type green melamine solid-top cap
closure type screw top vial
포장
pkg of 100 ea
O.D. × H
17 mm × 60 mm
용적
7 mL
피팅
thread for 15-425
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Vials are suitable to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Clear glass and amber glass offer essentially the same inertness.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)