주요 문서
Z740421
Tamper proof specimen containers
size 500 mL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.22
material
polypropylene cap
polypropylene container
포장
pack of 100 ea
제조업체/상표
Simport C581-500
크기
500 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. Tight-fitting lids prevent leaks and odors. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover. They resist temperatures up to 80 °C. The two larger sizes are supplied with a plastic handle for easy carrying.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)