주요 문서
Z329916
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 500 mL, label, Methanol, with venting valve (metal ball)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103209
NACRES:
NB.17
material
green closure
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
특징
label Methanol
with cap
with venting valve (metal ball)
포장
pack of 5 ea
제조업체/상표
BRAND 144026
용량
500 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
기타 정보
BR144026 (India)
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
부속물
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)