주요 문서
Z326380
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
volume 5 mL, accuracy: 0.04 mL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
material
borosilicate glass 3.3 flask
wide-neck
조인트
joint ST/NS neck10/19
설명
calibrated to contain (TC, IN)
특징
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
포장
pack of 2 ea
제조업체/상표
BRAND 37401
부피 측정 범위
5 mL, accuracy: 0.04 mL
용량
5 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The BLAUBRAND® volumetric flask combines precision and reliability, featuring a square grip PP stopper for easy handling. Crafted from high-quality Boro 3.3 glass and calibrated to contain (TC, In), each flask is marked with high-contrast white enamel for clear readability. Supplied with a batch number and certificate, these flasks ensure quality and traceability, making them ideal for laboratory use.
기타 정보
BR37401 (India)
법적 정보
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
프로토콜
This page contains detailed information on working with volumetric flasks and how to prepare a standard solution with a volumetric flask.