설명
with Sterile Glove
포장
pack of 100 ea
제조업체/상표
Nasco B01392WA
너비 × 길이 × 두께
11.5 cm × 23 cm × 0.064 mm
용량
532 mL × 18 oz
532 mL capacity(18 oz)
일반 설명
Includes a sterile polyethylene glove attached in a bottom compartment. To use the glove, tear off the glove pack along the perforation. The glove is folded with the open end slightly extended so the glove can be held and shaken open without touching the hand area.
Designed for environmental surface sampling of work areas, equipment, animal carcasses, and any other place where testing for Listeria, Salmonella, E.coli, coliforms, and other foodborne pathogens, or anthrax is required.
Designed for environmental surface sampling of work areas, equipment, animal carcasses, and any other place where testing for Listeria, Salmonella, E.coli, coliforms, and other foodborne pathogens, or anthrax is required.
법적 정보
Speci-Sponge is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
가장 최신 버전 중 하나를 선택하세요:
Global Trade Item Number
SKU | GTIN |
---|---|
WPB01392WA-100EA | 4061833576502 |