주요 문서
HS23412
Single Compartment Benchtop Dispensing Bin
blue
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.51
material
acrylic
blue
특징
holds Small laboratory consumables
포장
pack of 1 ea
제조업체/상표
Heathrow Scientific HS23412
길이 × 너비 × 높이
155 mm × 155 mm × 170 mm
일반 설명
The Benchtop dispensing bin works great with gloved hands and is ergonomically designed with large, angled openings and oversized lip.
특징 및 장점
  • Lid remains open when tilted back
  • Non-skid rubber feet and pre-drilled holes for wall mounting
  • Mounting screws included for the option of fixing to a wall or raised surface
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)