주요 문서
HS120568
Sprout® Plus Mini Centrifuge Portable Kit
AC/DC input 100 V / 240 V, universal plug set (plus 12V adapter)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
40161701
NACRES:
NB.11
특징
to hold 16 x 0.2 mL tubes
to hold 6 x 1.5/2 mL tubes
포장
pkg of 1 ea
제조업체/상표
Heathrow Scientific 120568
파라미터
2000 × g max. RCF (max.)
6,000 rpm speed (max.)
일반 설명
Spin Samples On-the-Go
The Portable Centrifuge Kit by Heathrow Scientific offers a convenient way to spin down and prepare samples outside of a conventional laboratory to prevent deterioration
Immediate Field Processing that is Easy to Use and offers Portable Convenience
The kit includes a Sprout Plus Mini Centrifuge securely housed in the DuraPorter transport box
Specially designed insert to seat the centrifuge, all its accessories and incorporates a 16-Place microtube rack. The DuraPorter is transparent for sample visibility and integrity checks
특징 및 장점
  • Tool free SnapSpin rotors
  • Quick-spin option
  • Includes 12V Car Adapter and Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs
  • DuraPorter® durably constructed of polycarbonate material
  • DuraPorter® has silicone water-tight seal and three lid clasps
  • Lockable with zip tie (not included)
포장
INCLUDES:
  • Sprout Mini Centrifuge
  • 6 Place 1.5/2.0 mL SnapSpin tube rotor
  • 16 Place 0.2 mL SnapSpin tube rotor (singles or 2x8 strip)
  • 6 Tube adapters (0.2 mL)
  • 6 Tube adapters (0.5 mL)
  • 4 O-rings
  • Storage case for rotors and adapters
  • 16 Place tube rack
  • Low voltage, double insulated power adapter with 4 interchangeable plugs
  • DuraPorter
  • Portable centrifuge case Inserts
  • 16 Place tube rack insert
  • Kit dividers
  • Sprout cradle
  • Portable 12-volt Sprout Centrifuge adapter
법적 정보
Duraporter is a registered trademark of Heathrow Scientific
Sprout is a registered trademark of Heathrow Scientific LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)