제조업체/상표
Heathrow Scientific HS120365
일반 설명
- Clock feature count up or down to 24 hours 59 minutes
- 4 timer channels
- Stands up or attaches to a magnetic surface (magnets on back of unit)
- Whiteboard and marking pen included
- Sounds when time expires
- Batteries included (2 AAA)
법적 정보
Lab Alert is a registered trademark of Heathrow Scientific
가장 최신 버전 중 하나를 선택하세요:
Global Trade Item Number
SKU | GTIN |
---|---|
HS120365 | 4061835296804 |
CRM45436-50MG | |
45436-250MG | 4061832340470 |