주요 문서
GE80-1129-32
GelBond Film 124 X 258 mm, (Pkg/50)
Cytiva 80-1129-32, pack of 50 pieces
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122410
NACRES:
NB.22
포장
pack of 50 pieces
제조업체/상표
Cytiva 80-1129-32
재고 정보
not available in Japan
저장 온도
room temp
일반 설명
GelBond PAGfilm is a polyester film with a treated surface which binds to the agarose gel.
저장 및 안정성
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
분석 메모
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Storage Class Code