주요 문서
GE28-9232-45
PD-10 Spin Adapter
Cytiva 28-9232-45, pack of 10 pieces
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115700
NACRES:
NA.56
특징
wetted part: no
포장
pack of 10 pieces
제조업체/상표
Cytiva 28-9232-45
일반 설명
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
특징 및 장점
  • Convenient adapters, enable spinning of all gravity columns.
Storage Class Code
13 - Non Combustible Solids
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
Lot/Batch Number
죄송합니다. 지금은 이 제품에 대한 COA이(가) 온라인에서 제공되지 않습니다.
문서
This page shows how to solve practical problems that may occur when running an affinity chromatography column.
프로토콜
This page shows how to prepare samples for purification with affinity chromatography.
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.