Key Documents
GE28-9232-43
MiniSpin Adapter
Cytiva 28-9232-43, pack of 10 pieces
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115700
NACRES:
NA.56
포장
pack of 10 pieces
제조업체/상표
Cytiva 28-9232-43
관련 카테고리
일반 설명
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
특징 및 장점
  • Convenient adapters, enable spinning of all gravity columns.
Storage Class Code
13 - Non Combustible Solids
문서 라이브러리 방문
프로토콜
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.