주요 문서
EPACSB-500S
Platform Clamps, for Eppendorf® Shakers
for Eppendorf® New Brunswick Innova 2000 shaker, for 500 mL media bottles
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103816
NACRES:
NB.73
특징
for 500 mL media bottles
for Eppendorf® New Brunswick Innova 2000 shaker
제조업체/상표
Eppendorf® ACSB-500S (Global)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Flask clamps are of a single piece stainless-steel construction (not spot welded or riveted) for added strength and safety. Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds. Flask clamps come complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
법적 정보
Eppendorf is a registered trademark of Eppendorf AG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)