Key Documents
EP5820774002
Eppendorf® Steel Sleeves
for rotor F-35-48-17, F-48x15, set of 24 pieces
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103911
NACRES:
NB.82
특징
Holds 15 mL tubes
for rotor F-35-48-17, F-48x15
포장
set of 24 pieces
제조업체/상표
Eppendorf® 5820774002
기술
centrifugation: suitable
호환성
for use with F-48x15
for use with Rotor F-35-48-17
법적 정보
Eppendorf is a registered trademark of Eppendorf AG