주요 문서
EP5490042000
Rotor for Eppendorf® Concentrator Plus
F-50-8-18, non aerosol tight, to hold, 8 x 10/18 mL round-bottome tubes
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103909
NACRES:
NB.82
특징
to hold 8 x 10/18 mL round-bottome tubes
to hold 8 x 18 x 105 - 128 mm
fixed angle rotor
non aerosol tight
포장
pack of 1 ea
제조업체/상표
Eppendorf® 5490042000 (EU)
기술
centrifugation: suitable
호환성
for use with centrifuge models Concentrator Plus
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Fixed angle rotor F-50-8-18 can hold a maximum of 8 x 10/18 mL round-bottomed tubes with diameter of 105 - 128 mm. It is not aerosol tight and is to be used with Eppendorf Concentrator Plus.
Selection of rotors for the Eppendorf Concentrator Plus.
법적 정보
Eppendorf is a registered trademark of Eppendorf AG