주요 문서
EP0030131525
Eppendorf® plate lid
sterile, pkg of 80 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.22
무균
sterile
포장
pkg of 80 ea
제조업체/상표
Eppendorf® 0030131525
유사한 제품을 찾으십니까? 방문 제품 비교 안내
특징 및 장점
  • Sealing Mats are reusables and can be autoclave. Sealing mats are suitable for short-term storage
    • Heat Sealing Film and Foil for continuous locking give the best evaporation protection
    • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
    • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
    • Storage Film and Foil glued seals for simple and dependable sealing during sample storage
법적 정보
Eppendorf is a registered trademark of Eppendorf AG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)