주요 문서
EL1
Sigma-Aldrich
Endura-Luers
Stopcocks for vacuum manifold
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41105501
NACRES:
NA.52
Quality Level
일반 설명
Sigma offers Endura-Luer stopcocks to be used in conjunction with the VM20 Vacuum Manifold. As an alternative to plugs, they create maximum efficiency when purifying multiple preps. Endura-Luers resist chemical erosion, cracking, and breaking.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)