주요 문서
ZWATSENA1
Milli-Q
Water sensor
Safety accessory that detects a water leak, for use with Milli-Q® IQ/IX/EQ/SQ series.
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104200
NACRES:
JA.13
AC/DC 입력
120 V, 60 Hz
포장
pkg of 1 unit
호환성
for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® SQ 2Series
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
  • The Water sensor is a safety accessory which detects the presence of excess water on the floor and automatically shuts down the feed water access from the water system or main water supply.
  • It is usually placed on the floor or a benchtop, wherever is most convenient for the lab.
  • The sensor has a 4 m cable so can be connected to the water purification system up to 4 m away.
애플리케이션
This water sensor accessory for Milli-Q® IQ/IX/EQ/SQ series water purification systems prevents water damage to the lab, in an unlikely event of a leak.
특징 및 장점
  • Highly reduces the risk of flooding.
  • Protects the lab instruments and surroundings from water damage.
법적 정보
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
문서 라이브러리 방문
프로토콜
Milli-Q® SQ 2Series 실험용 정제수 시스템에 대한 지침 영상을 보고 사용자 설명서를 다운로드하십시오. 즉시 설치하여 사용할 수 있습니다. 시스템, 액세서리, 카트리지 및 서비스를 온라인으로 구매하십시오.