주요 문서
ZFMQPR3BT0
Milli-Q
Milli-Q® Pressure Regulator
Limits tap feed water pressure to 3 bars to ensure optimal RO membrane performance
로그인조직 및 계약 가격 보기
크기 선택
크기 선택
보기 변경
About This Item
UNSPSC 코드:
41104200
eCl@ss:
33050190
NACRES:
NC.75
포장
pkg of 1 unit
일반 설명
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The Milli-Q® Pressure Regulator is dedicated to limit tap feed water pressure to 3 Bars, to ensure optimal RO membrane performance of Milli-Q® systems.
It needs to be connected to the feed water tap, just before the water purification system and can be wall mounted.
Compatible with all systems from the Milli-Q® SQ 2Series family with an RO production station.
법적 정보
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany