주요 문서
SYMBLTVP1
Millipore
Steritest® Symbio Pumps Validation Protoco
Letter format
로그인조직 및 계약 가격 보기
크기 선택
크기 선택
보기 변경
About This Item
UNSPSC 코드:
41116500
NACRES:
JA.11
Quality Level
호환성
for use with Steritest® Symbio Pumps
일반 설명
Steritest® Symbio Pumps validation protocol is designed to make it easier for the technicians to ensure compliance of the symbio pumps for consistent performance. The product is GMP compliant and includes Validation Master Plan (VMP), Installation Qualification (IQ), Operational Qualification (OQ) and Performance Qualification (PQ) test protocols. In addition to the above, ready to be filled report table formats are also provided.
애플리케이션
Steritest® Symbio Pumps validation protocol is a ready to use GMP compliant validation protocol along with ready to be filled report table formats.
법적 정보
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany