주요 문서
RDHSTAND1
Milli-Q
Stand for Remote Dispenser
For Direct-Q® and Synergy® remote systems
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104200
NACRES:
SB.51
포장
pkg of 1 unit
호환성
for use with Direct-Q® UV
for use with Direct-Q®
for use with Synergy® UV
for use with Synergy®
일반 설명
The versatile remote dispenser, when supported by this stand, can be placed on a bench up to 2 m from the water purification unit.
애플리케이션
For Direct-Q® or Synergy® remote systems.Provides the support to place the Remote Dispenser on a lab bench.
법적 정보
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany