주요 문서
Z743166
Julabo® bath covers
flat cover to fit CORIO CD-B33
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103706
NACRES:
NB.77
포장
pack of 1 ea
제조업체/상표
Julabo 9970292
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Flat stainless steel bath cover reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath.
법적 정보
Julabo is a registered trademark of Julabo USA, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)