추천 제품
AC/DC 입력
5 - 400 mA (Max Wattage: 80W; Voltage Output: 10-300V (5V increments))
특징
output terminals: 4 pairs
touchscreen control; Dual orientation, auto rotating display
포장
pkg of 1
제조업체/상표
Benchmark Scientific E2301
호환성
with most brands of gel boxes
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Accuris myVolt Touch is a dependable electrophoresis power supply that includes all of the performance and specifications needed for efficient seperations of nucleic acids and proteins. Perfect for agarose gels and protein seperations, the myVolt Touch incorporates some very unique user-friendly features: Touch-screen Control Panel: Setting up an electrophoresis run with the myVolt Touch couldn′t be easier... simply touch the parameter that you want to change and then touch "start" to begin the run.
Two sets of output jacks are available to connect up to 2 gel rigs and the instruments can securely be stacked for use with additional gel rigs.
Two sets of output jacks are available to connect up to 2 gel rigs and the instruments can securely be stacked for use with additional gel rigs.
법적 정보
myVolt is a trademark of Benchmark Scientific, Inc.
가장 최신 버전 중 하나를 선택하세요:
Global Trade Item Number
SKU | GTIN |
---|---|
Z742604-1EA | 4061826162132 |