특징
12-channel
color-coded yellow
electronic: no
포장
pack of 1 ea
제조업체/상표
Finn 4662060
용적
10-100 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Double action pipetting button with very light and smooth pipetting action
- Fully autoclavable
- Super blow-out piston with volumes of 50 μl and below
법적 정보
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
가장 최신 버전 중 하나를 선택하세요:
Global Trade Item Number
SKU | GTIN |
---|---|
Z678058-1EA | 4061833035900 |