제조업체/상표
Plas-Labs 890-THC/DT/EXP/EU
내부 높이(H) × 너비(W) × 직경(D)
26 in. × 41 in. × 28 in.
외부 높이(H) × 너비(W) × 직경(D)
38 in. × 55 in. × 35 in.
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Price includes shipping
The Plas-Labs temperature and humidity control glove box is designed to maintain accurate control of the isolated atmosphere. The drying train can be used when in-house inert gas is not available. The molecular sieve should be regenerated periodically for optimal drying performance.
The easy to use touch screen offers password protection, on screen readings of chamber temperature and humidity, full color screen, and room temperature and humidity readings.
The easy to use touch screen offers password protection, on screen readings of chamber temperature and humidity, full color screen, and room temperature and humidity readings.
- Humidity (Rh) transmitter with 8ft. sensor cable
- Two inlet/outlet gas valves on main chamber and transfer chamber
- Serial port connection for data logging
- Thermoelectric assembly (Peltier device)
- Nebulizer
- Two drying train canisters (filled with Molecular Sieve) and circulation pump
- CE compliant
법적 정보
관련 제품
제품 번호
설명
가격
가장 최신 버전 중 하나를 선택하세요:
Global Trade Item Number
SKU | GTIN |
---|---|
Z660612-1EA | 4061833574256 |