material
polycarbonate
특징
Can be stacked three high.
포장
pack of 1 ea
제조업체/상표
model 1.0
Bel-Art F42071-0008
높이 × 너비 × 깊이
21.3 cm × 34.1 cm × 41.4 cm
문 개방부 높이 × 너비
11.4 cm × 22.5 cm
색상
amber
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Dry storage of humidity sensitive products is now easier. Secador cabinets assure moisture and dust free storage of valuable items like reagents and pipettes with their proprietary sealed design.
Available in small or large models, with either horizontal or vertical orientation, these uniquely-designed units are available totally clear, clear with blue end caps, and amber. The stackable small model is perfect for maximizing storage space in a minimum footprint.
Cabinets are molded of polycarbonate resistant to staining, crazing and chemical attack and have built-in protection that blocks 99% of UV light. Desiccants can be placed under the bottom shelf or on any shelf. A hygrometer in the door allows for easy monitoring of relative humidity. Carrying handles make them easy to relocate.
Amber cabinets reduce visible light penetration by more than 50% and blocks 99% of UV light.
Available in small or large models, with either horizontal or vertical orientation, these uniquely-designed units are available totally clear, clear with blue end caps, and amber. The stackable small model is perfect for maximizing storage space in a minimum footprint.
Cabinets are molded of polycarbonate resistant to staining, crazing and chemical attack and have built-in protection that blocks 99% of UV light. Desiccants can be placed under the bottom shelf or on any shelf. A hygrometer in the door allows for easy monitoring of relative humidity. Carrying handles make them easy to relocate.
Amber cabinets reduce visible light penetration by more than 50% and blocks 99% of UV light.
- Has two perforated shelves that are removable
- Door has two latches at the top and loops for - lock or tamper evident seal
기타 정보
Model 1.0 has 1 fixed shelf and 1 door latch. Models 3.0 and 4.0 have 3 removable shelves.
법적 정보
Scienceware is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
이미 열람한 고객
Global Trade Item Number
SKU | GTIN |
---|---|
Z566756-1EA | 4061832265872 |