주요 문서
Z231320
Sigma-Aldrich
Giant-size molecular model kit
deluxe kit
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
60104506
NACRES:
NB.17
설명
no. of flexible bonds 32
no. of rigid bonds 100
특징
no. of atoms 64
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Large, lightweight molecular models for classroom demonstration of organic and inorganic structures. For building models to scale of 20 cm/Å and 8 cm/Å. Bond lengths range from 5.5 to 27 cm. Atomic centers are 5 cm in diameter, finished in standard atom colors, and have a reinforced vinyl receptor at each bond position for insertion of connecting rods (bonds). All model kits include storage box and instructions. Additional atoms and bonds are available separately.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)