특징 및 장점
- Provided with a 4mm polytetrafluoroethylene (PTFE) stopcock
- Has no frit and uses a glass-wool plug and sand
- Use along with coarse silica gel (e.g., Cat. No. 60744) for fast and general-purpose purification.
가장 최신 버전 중 하나를 선택하세요:
이미 열람한 고객
Global Trade Item Number
SKU | GTIN |
---|---|
Z164003-1EA | 4061837606687 |