주요 문서
Z108898
Sigma-Aldrich
Mini Diazald® apparatus
joint: ST/NS 19/22 Clear-Seal joints, for generating 1 to 50 mmol of diazomethane
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104812
NACRES:
NB.43
조인트
joint: ST/NS19/22 Clear-Seal joints
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
For technical and safety information about Diazald and Aldrich diazomethane generators see Technical Information Bulletin AL-180.
The mini Diazald apparatus bridges the gap between the System 45 diazomethane generator Z411736 (for preparing <1 mmol of diazomethane) and the Diazald kit (for preparing ~100 mmol of diazomethane). It consists of a reaction vessel and a condenser in one compact unit. This apparatus features a cold-finger in place of a water-jacketed condenser. When filled with dry ice/acetone slush, the condenser efficiently prevents dangerous diazomethane/ether vapor from escaping. A separatory funnel, Z100382 and a round-bottom receiving flask, Z100358, (must be equipped with Clear-Seal joints) are the only other glassware needed and can be purchased separately.
NOTE: Sigma-Aldrich is unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
법적 정보
Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Lalit K Golani et al.
Journal of medicinal chemistry, 59(17), 7856-7876 (2016-07-28)
Targeted antifolates with heteroatom replacements of the carbon vicinal to the phenyl ring in 1 by N (4), O (8), or S (9), or with N-substituted formyl (5), acetyl (6), or trifluoroacetyl (7) moieties, were synthesized and tested for selective
Global Trade Item Number
SKUGTIN
Z108898-1EA4061832114576