주요 문서
T289604
(2S,3AS,5AR,5BS,9S,13S,14R,16AS,16BS)-9-ETHYL-13-HYDROXY-14-METHYL-7,15-DIOXO-2,3,3A,5A,5B,6,7,9,10,11,12,13,14,15,15A,16,16A,16B-OCTADECAHYDRO-1H-AS-INDACENO[3,2-D]OXACYCLODODECIN-2-YL 6-DEOXY-2,3,4-TRI-O-METHYL-ALPHA-L-MANNOPYRANOSIDE
AldrichCPR
로그인조직 및 계약 가격 보기
About This Item
Linear Formula:
C33H52O9
Molecular Weight:
592.777
MDL number:
기타 정보
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)