주요 문서
MT51344809
Mettler-Toledo perfectION combination electrode
Measurement range 8 * 10-6-10-2 mol/L & 0.2-260 mg/L
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
39121436
NACRES:
NB.79
material
epoxy shaft
glass shaft (solid state)
설명
Lemo connector
제조업체/상표
Mettler-Toledo 51344809
케이블 크기
1.2 m
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Mettler Toledo perfectION ISE Combined ion-selective electrode means ISE measurement has never been so simple!
The new Click & Clear junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
  • Sensor with attached cable and BNC connector
  • Reference electrolyte (60 mL)
  • Operating instructions
    To ensure an unproblematic start to your ISE measurement, the required calibration standards and ISA solutions (Ionic Strength Adjuster) can be ordered at the same time.
comb CN Lemo
법적 정보
Click & Clear is a trademark of Mettler-Toledo, Inc.
perfectION is a trademark of Mettler-Toledo, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)