주요 문서
MT51343174
Mettler Toledo InLab® Cool pH electrode
for use in low temperature samples
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115604
NACRES:
NB.79
material
glass (LoT)
glass membrane (polymer)
설명
S7 connector
without cable
제조업체/상표
Mettler-Toledo 51343174
길이 × 너비
120 mm × 12 mm
일반 설명
The Mettler Toledo InLab® Cool is a glass-body, combination pH electrode with S7 screw head. Reference electrode is filled with Friscolyte-B electrolyte for low temperature samples.
By filling the reference electrode with Friscolyte-B® electrolyte the electrode can be used in samples down to a temperature of minus thirty degrees Centigrade.
ARGENTHAL reference system improves service life and measurement accuracy at differing sample temperatures.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • ARGENTHAL reference
  • Friscolyte-B electrolyte
  • Detachable cable
법적 정보
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)