주요 문서
MT51343056
Mettler-Toledo InLab® Routine & Routine Pro pH electrodes
L 120 mm × 12 mm, MultiPin connector, Temperature probe Pt1000
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115604
NACRES:
NB.79
material
glass shaft
glass, HA membrane
설명
Without cable
제조업체/상표
Mettler-Toledo 51343056
온도 측정 범위
0-100 °C
길이
120 mm × 12 mm
pH
0-14
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
InLab® Routine are glass-body, combination pH electrodes with S7 screw head.
Designed for accurate measurement in a wide range of sample types.
The electrolyte in conventional silver/silver chloride reference systems must be saturated with silver chloride. With critical media such as those containing sulphides or proteins, this will result in diaphragm contamination with silver sulphide. This cannot happen with InLab electrodes which have an ARGENTHAL lead off and silver ion trap that ensure the electrolyte remains completely free from silver ions.
The ARGENTHAL reference system also improves service life and measurement accuracy at differing sample temperatures. The glass shaft provides resistance against organic and corrosive samples.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • ARGENTHAL reference
  • Silver ion trap
  • Glass shaft
  • Detachable cable
Routine Pt1000
법적 정보
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
MultiPin is a trademark of Mettler-Toledo, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)