material
borosilicate glass bottle
cylindrical bottle
phenolic cap
특징
autoclavable
cap
graduations
포장
pack of 12 ea
case of 48 ea
제조업체/상표
Corning 1373-160
bottle neck diam.
~72 mm
용량
160 mL
일반 설명
Wide mouth bottle for easy measuring or filling. With cut-line graduation mark at 99 ± 1mL. Phenolic cap with rubber liner will withstand sterilization (wet at 121°C).
법적 정보
Corning is a registered trademark of Corning, Inc.
가장 최신 버전 중 하나를 선택하세요:
Global Trade Item Number
SKU | GTIN |
---|---|
CLS1373160-12EA | 4061833881071 |
CLS1373160-48EA | 4061833881088 |