주요 문서
BAF424002221
Scienceware® Lab Companion round vacuum desiccator
volume 20 L, clear polycarbonate, pack of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104510
NACRES:
NB.43
material
clear polycarbonate
포장
pack of 1 ea
제조업체/상표
Bel-Art F42400-2221
용적
20 L
20000 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
These high-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind. Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods. A Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange.
Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
Available in both cabinet and round styles, all models include a desiccant tray (desiccants sold separately).
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • Locking ring secures top and bottom halves when vacuum not in use; provides additional convenience and safety when moving the unit
  • Six models to choose from; 6, 10, and 20 L internal volume in clear or UV blocking amber
법적 정보
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)