주요 문서
BAB609011300
SP Bel-Art® H-B Durac® Thermocouple Thermometer Probe
K-TYPE
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41112219
NACRES:
NB.79
포장
pack of 1 ea
제조업체/상표
HB Instruments B60901-1300
일반 설명
For use with H-B DURAC Calibrated Thermocouple Thermometers or others that accept K or J type probes.
특징 및 장점
  • For temperature measurement of liquids, semi-solids, or air/gasses
  • Measure wide temperature ranges
  • 15.24cm (6”) stainless steel probe
  • B60901-1300 perfect for use in dewars
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)