Skip to Content
Merck
All Photos(1)

Documents

MIRRM00

Sigma-Aldrich

MystiCq® microRNA® SYBR® Green qPCR ReadyMix

Formulated for systems that do not require normalization

Synonym(s):

SYBR® Green-based miRNA qPCR master mix, miRNA qPCR, microRNA qPCR master mix without ROX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

technique(s)

qPCR: suitable

compatibility

Bio-Rad CFX384
Bio-Rad CFX96
Bio-Rad MJ Chromo4
Bio-Rad MJ Opticon 2
Bio-Rad MJ Opticon
Bio-Rad MiniOpticon
Cepheid SmartCycler
Eppendorf® Mastercycler ep realplex2 s
Eppendorf® Mastercycler ep realplex
Illumina Eco qPCR
Qiagen Corbett Rotor-Gene 3000
Qiagen Corbett Rotor-Gene 6000
Qiagen Corbett Rotor-Gene Q
Roche LightCycler 480

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The MystiCq® microRNA SYBR® Green qPCR ReadyMix is one of the integral components of the MystiCq microRNA qPCR Assay System. This master mix is a specially formulated 2X concentrate with an antibody-based hot-start Taq DNA polymerase and all the optimized components needed for highly specific, reliable, and reproducible microRNA qPCR assays. Simply add the specific MystiCq microRNA qPCR Assay Primer, the MystiCq Universal PCR primer, and cDNA template generated by the MystiCq microRNA cDNA Synthesis Kit and you′re ready to go. Because there are different methods used for fluorescent normalization on different real-time PCR instruments, there is a MystiCq microRNA SYBR Green qPCR ReadyMix available for your platform.

Application

MystiCq microRNA SYBR Green qPCR ReadyMix is compatible with the following platforms: Bio-Rad CFX384, Bio-Rad CFX96, Bio-Rad MiniOpticon, Bio-Rad/MJ Chromo4, Bio-Rad/MJ Opticon 2, Bio-Rad/MJ Opticon, Cepheid SmartCycler®, Eppendorf Mastercycler® ep realplex, Eppendorf Mastercycler ep realplex2 s, Illumina Eco qPCR, Qiagen/Corbett Rotor-Gene® 3000, Qiagen/Corbett Rotor-Gene 6000, Qiagen/Corbett Rotor-Gene Q, Roche LightCycler® 480.

Features and Benefits

  • Exclusively for use with the MystiCq microRNA qPCR Assay Primers, Control Primers, and Universal PCR primer
  • 2X concentrated for easy reaction setup with hot-start Taq, dNTPs and optimized buffer
  • Passive reference dye (if required) included at the optimal concentration
  • This master mix can readily amplify fragments between 400 and 500 bp; however, for best results, amplicon size should be limited to 80 - 200 bp

Packaging

Default reaction volume is 50 μL

100RXN is packaged as 2 X 1.25 mL
500RXN is packaged as 10 X 1.25 mL
2000RXN is packaged as 1 X 50 mL

Legal Information

CFX384 is a trademark of Bio-Rad Laboratories, Inc.
CFX96 is a trademark of Bio-Rad Laboratories, Inc.
Chromo4 is a trademark of Bio-Rad Laboratories, Inc.
Eppendorf is a registered trademark of Eppendorf AG
LightCycler is a registered trademark of Roche
Mastercycler is a registered trademark of Eppendorf AG
MystiCq is a registered trademark of QIAGEN Beverly Inc.
Opticon is a trademark of Bio-Rad Laboratories, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
Rotor-Gene is a registered trademark of Qiagen GmbH
SYBR is a registered trademark of Life Technologies
SmartCycler is a registered trademark of Cepheid, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Baccelli Mendonça et al.
Heliyon, 6(12), e05734-e05734 (2020-12-29)
Regulation of mTOR signaling depends on an intricate interplay of post-translational protein modification. Recently, mEAK-7 (mTOR associated protein, eak-7 homolog) was identified as a positive activator of mTOR signaling via an alternative mTOR complex. However, the upstream regulation of mEAK-7
Katarzyna Goljanek-Whysall et al.
Development (Cambridge, England), 141(17), 3378-3387 (2014-08-01)
Myogenesis involves the stable commitment of progenitor cells followed by the execution of myogenic differentiation, processes that are coordinated by myogenic regulatory factors, microRNAs and BAF chromatin remodeling complexes. BAF60a, BAF60b and BAF60c are structural subunits of the BAF complex
A Mukherjee et al.
Andrology, 3(5), 979-990 (2015-08-28)
Mechanistic target of rapamycin (mTOR) is a signal transduction pathway that modulates translation initiation in several animals including mammals. Rapamaycin, an allosteric inhibitor of mTOR pathway, is often used as an immunosuppressive drug following kidney transplantation and causes gonadal dysfunction
Guillaume Vares et al.
PloS one, 9(8), e106277-e106277 (2014-08-30)
Both exposure to ionizing radiation and obesity have been associated with various pathologies including cancer. There is a crucial need in better understanding the interactions between ionizing radiation effects (especially at low doses) and other risk factors, such as obesity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service