Skip to Content
Merck
All Photos(1)

Key Documents

B4187

Sigma-Aldrich

BRK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PTK6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

113-153 nmol/min·mg

mol wt

~80 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTK6(5753)

Biochem/physiol Actions

BRK is a member of the non-receptor tyrosine kinases (PTKs) that contains an amino terminal SH3 and SH2 domain as well as the catalytic domain. BRK expression is low or undetectable in normal mammary tissue and benign lesions. However, approximately two-thirds of breast tumors express appreciable levels and 27% of tumors over express BRK by fivefold or more.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Mitchell et al.
Oncogene, 15(12), 1497-1502 (1997-10-23)
The brk gene encodes a non-receptor protein tyrosine kinase that consists of single SH3, SH2 and catalytic domains. Although BRK shows strongest sequence similarity to members of the SRC family of PTKs, there are several key structural and regulatory differences
P J Mitchell et al.
Oncogene, 9(8), 2383-2390 (1994-08-01)
Using a polymerase chain reaction based differential screening approach, we have isolated and characterised a cDNA from a human metastatic breast tumour representing a novel protein tyrosine kinase (brk). Sequencing of brk cDNAs isolated from T-47D and MCF-7 human breast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service