IPAKQUAA1
IPAK Quanta® Polishing Cartridge
Designed for use with Milli-Q® IQ 7 and EQ 7 series water purification systems
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
shipped in
ambient
Related Categories
General description
IPAK Quanta® polishing cartridges are lab water purification consumables designed for use with Milli-Q® 7000/03/05/10/15 and Milli-Q® EQ 7000/08/16 water purification systems. These systems deliver superior-quality ultrapure water for sensitive research applications.
IPAK Quanta® polishing cartridge contains innovative IQnano™ ion-exchange media which achieves ion removal down to trace levels. The small bead size significantly improves kinetic properties while dramatically reducing media volume — 33% less than former Milli-Q® purification cartridges. In addition, high-grade synthetic activated carbon targets trace organic contaminants.
For Milli-Q® IQ 7000 and Milli-Q® EQ 7000/08/16 water purification systems, the IPAK Meta® and IPAK Quanta® cartridges work as a pair to produce highest-quality ultrapure water.
IPAK Quanta® polishing cartridge contains innovative IQnano™ ion-exchange media which achieves ion removal down to trace levels. The small bead size significantly improves kinetic properties while dramatically reducing media volume — 33% less than former Milli-Q® purification cartridges. In addition, high-grade synthetic activated carbon targets trace organic contaminants.
For Milli-Q® IQ 7000 and Milli-Q® EQ 7000/08/16 water purification systems, the IPAK Meta® and IPAK Quanta® cartridges work as a pair to produce highest-quality ultrapure water.
Features and Benefits
- A full report is available for the IPAK Quanta® cartridge. It provides details on the technologies used, their performance during testing by our R&D department, as well as information concerning our quality and manufacturing requirements.
- e-Sure tag for RFID connection enables full traceability (data management) and automatic consumable status monitoring on the POD’s touchscreen interface.
- 78% of the parts suppliers are ISO 14001 environmental management system-certified.
Legal Information
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
also commonly purchased with this product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service