Skip to Content
Merck
All Photos(2)

Key Documents

525162

Sigma-Aldrich

Sucrose diacetate hexaisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H62O19
Molecular Weight:
846.91
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

optical activity

[α]20/D +49°, c = 1 in chloroform

refractive index

n20/D 1.457 (lit.)

bp

288 °C (lit.)

mp

−7 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(=O)OC[C@H]1O[C@H](O[C@@]2(COC(C)=O)O[C@H](COC(=O)C(C)C)[C@@H](OC(=O)C(C)C)[C@@H]2OC(=O)C(C)C)[C@H](OC(C)=O)[C@@H](OC(=O)C(C)C)[C@@H]1OC(=O)C(C)C

InChI

1S/C40H62O19/c1-18(2)33(43)51-17-40(32(57-38(48)23(11)12)29(54-35(45)20(5)6)27(58-40)16-50-25(14)42)59-39-31(56-37(47)22(9)10)30(55-36(46)21(7)8)28(53-34(44)19(3)4)26(52-39)15-49-24(13)41/h18-23,26-32,39H,15-17H2,1-14H3/t26-,27-,28-,29-,30+,31-,32+,39-,40+/m1/s1

InChI key

ZNEBZIJCDDCNRC-SWTLDUCYSA-N

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rasmus I Jølck et al.
Advanced healthcare materials, 3(10), 1680-1687 (2014-04-16)
External beam radiation therapy relies on the ability to deliver high radiation doses to tumor cells with minimal exposure to surrounding healthy tissue. Advanced irradiation techniques, including image-guided radiation therapy (IGRT), rely on the ability to locate tumors to optimize
Jianbo Guo et al.
AAPS PharmSciTech, 16(4), 973-985 (2015-01-23)
Our previous studies revealed that the PLGA-based particulate systems loaded with cucurbitacin showed limited anti-melanoma efficiency in xenograft animal models after intratumoral injection, which was due to the undesirable initial burst release and the leakage of the particulate carriers from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service