Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-tri-Calcium phosphate
Synonym(s):
β-TCP, β-Tricalcium phosphate
Linear Formula:
Ca
3
(PO
4
)
2
CAS Number:
7758-87-4
Molecular Weight:
310.18
EC Number:
231-840-8
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Purity
Quality Segment
Particle Size (µm)
Special Grade
Puriss (2)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
49963
β-tri-Calcium phosphate,
puriss. p.a., ≥98% β-phase basis (sintered Powder)
Expand
View Pricing
900205
β-tri-Calcium phosphate,
powder, 4 μm, ≥80 m
2
g
Expand
View Pricing
13204
β-tri-Calcium phosphate,
puriss. p.a., ≥98% β-phase basis (unsintered powder)
Expand
View Pricing