Ethylenediaminetetraacetic acid
Synonym(s):
(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid, OmniPur® EDTA, Titriplex® II, Ethylenediaminetetraacetic acid
Linear Formula:
(HOOCCH2)2NCH2CH2N(CH2COOH)2
CAS Number:
Molecular Weight:
292.24
Products
Shipping
Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing