4008-M
Ethylenediaminetetraacetic acid
OmniPur® Grade, >= 99.5%
Synonym(s):
OmniPur® EDTA, (Ethylenedinitrilo)tetraacetic Acid, Ethylenediaminetetraacetic Acid
About This Item
Recommended Products
grade
reagent
Assay
≥99.5%
form
crystalline powder (Fine)
storage condition
OK to freeze
color
white to off-white
cation traces
Fe: ≤0.005%
Mg: ≤0.0005%
heavy metals (as Pb): ≤0.001%
shipped in
ambient
InChI
1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)
InChI key
KCXVZYZYPLLWCC-UHFFFAOYSA-N
General description
Ash: ≤0.2%
Assay: ≥99.5%
Color of 0.5M Solution (APHA): ≤30
DNAse: none detected
Heavy metals (as Pb): ≤0.001%
Iron (Fe): ≤0.005%
Magnesium (Mg): ≤0.0005%
Nitrilotriacetic acid: ≤0.1%
pH (5%, water): 2.5-3.5
Protease: none detected
RNAse: none detected
Solubility (5%, 1 N NaOH): To pass test
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service