Merck
HomeProductsLabware & Water PurificationWater PurificationMilli-Q® Benchtop Water Purification Systems
Milli-Q® Benchtop Water Purification Systems
Type 1 and Type 2 water purification system with production and tank units plus pure and ultrapure water dispensers. A combination of purification technologies: pre-filtration, reverse osmosis, electrodeionization, UV, and final filtration.
Our Milli-Q® Lab Water Solutions portfolio of benchtop water systems includes both pure and ultrapure systems to meet any lab’s needs. From sensitive QC analyses to buffer preparation and glassware rinsing, this collection of compact systems has your lab covered. Our specialists can help you find the water purification solution that’s best adapted to your requirements in terms of:
  • Applications
  • Water quality grade
  • Volume demand
  • Lab configuration
  • Budget
Choosing the Correct Water Quality for your Applications
Water grades in the lab are defined by quality standards, such as the ASTM or ISO 3696, which help ensure that the correct water quality is used for specific applications. Standards also consider the cost of production, as Type 1 ultrapure water is more expensive to produce than Type 2 or Type 3 pure water.
Type 3 water is the lowest water grade and is produced by reverse osmosis (RO). RO water is recommended for non-critical lab applications, such as glassware rinsing, heating baths, autoclaves, and as feed for Type 1 water systems.
Type 2 pure water is recommended for general lab use, such as to prepare buffers, reagents, and culture media, and as feed for other equipment (e.g. Type 1 systems, clinical analyzers, dishwashers, stability test chambers, etc.). Our Type 2 systems incorporate environmentally friendly Elix® electrodeionization (EDI) technology which produces constant-quality pure water while eliminating the need for hazardous chemical regeneration procedures, costly resins, changing DI cartridges, and adding softeners.
Type 1 ultrapure water is ion-free (resistivity 18.2 MOhm·cm) with low total organic carbon (TOC) content. Ultrapure water is recommended for sensitive laboratory applications such as advanced analytical techniques (e.g. HPLC, UHPLC, LC-MS, ICP-MS, GF-AAS), molecular biology applications (e.g. PCR, sequencing), and to prepare buffers and culture media for mammalian cell culture.
Space-saving and Flexible Options—From Installation to Dispensing
Our benchtop systems are designed to save valuable lab space, providing versatile installation, storage, and dispensing options. Our most recent Milli-Q® IQ and IX series systems require only the slim POD dispenser on the bench. The compact purification unit and tank can be integrated under the bench, on the wall, or a combination of both to suit your requirements.
For labs needing more than one point-of-use, multiple dispensers can be connected to one system, with up to 5 m between the PODs. This way, scientists at distant benches—or even another lab—have access to purified water. In addition, a range of production flow rates and tank size options are available to ensure your lab's needs are fully met.
Digital Services to Simplify System Care
MyMilli-Q™ Online Solution is a unique digital service that simplifies the care of your Milli-Q® water system. Here are some of the tasks you can accomplish by simply logging into your MyMilli-Q™ account:
  • Track service history
  • Download service reports
  • Schedule maintenance visits
  • Track consumable orders
  • Manage consumable deliveries
  • View contact details
  • Renew your service plan
  • Contact Milli-Q® Services
With these time-saving features, you’ll benefit from:
  • At-a-glance, consolidated contract information
  • More rapid and efficient planning
  • Streamlined audit preparation
  • Easily traceable service history and reports
MyMilli-Q™ Online Solution is available for all systems under warranty or with a Milli-Q® Service Plan.
Highlights