Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

V800175

Sigma-Aldrich

Ethanolamine

LR, ≥99%

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

LR

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

autoignition temp.

1436 °F

expl. lim.

17 %

refractive index

n20/D 1.454 (lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water soluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

V800175-VAR:
V800175-500ML-PW:
V800175-4X2.5L:4548173328218
V800175-2.5L-PW:
V800175-6X500ML:4548173328225
V800175-BULK:
V800175-500ML:4548173324067
V800175-2.5L:4548173324050


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Vance
Biochimica et biophysica acta, 1045(2), 128-134 (1990-07-16)
Monolayer cultures of rat hepatocytes have been examined for their ability to secrete ethanolamine plasmalogen as a component of nascent lipoproteins. In culture medium from these cells, ethanolamine plasmalogen comprises approx. 20-30% of total ethanolamine glycerophospholipids when measured either as
H Perschak et al.
Human neurobiology, 6(3), 191-194 (1987-01-01)
Concentrations of putative neuroactive substances glutamate, aspartate, gamma-aminobutyric acid, glycine, proline and ethanolamine were determined in ventricular cerebrospinal fluid collected in patients suffering from Parkinson's disease, pain syndromes or cerebellar tremor. Values are similar to those given in the literature
Gabriel da Silva
The journal of physical chemistry. A, 116(45), 10980-10986 (2012-09-22)
The alkanolamine 2-aminoethanol (NH(2)CH(2)CH(2)OH), otherwise known as monoethanolamine (MEA), is a widely used solvent for carbon capture, yet relatively little is known about its atmospheric chemistry. The hydroxyl radical initiated oxidation of MEA is thought to predominantly form the α-aminoalkyl
Danielle A Garsin
Nature reviews. Microbiology, 8(4), 290-295 (2010-03-18)
Ethanolamine is a compound that can be readily derived from cell membranes and that some bacteria can use as a source of carbon and/or nitrogen. The complex biology and chemistry of this process has been under investigation since the 1970s
Ye-Jin Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(31), 4578-4584 (2015-07-03)
By controlling the polymer/polymer blend self-organization rate, all-polymer solar cells composed of a high-mobility, crystalline, naphthalene diimide-selenophene copolymer acceptor and a benzodithiophene-thieno[3,4-b]thiophene copolymer donor are achieved with a record 7.7% power conversion efficiency and a record short-circuit current density (18.8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service