501670
Carbon Monoxide, Carbon Dioxide, Hydrogen, Methane and Oxygen
1 mol each component in nitrogen, aerosol can of 4 L, analytical standard
About This Item
grade
analytical standard
form
gas
packaging
aerosol can of 4 L
manufacturer/tradename
Scott Gas T218-4
concentration
1 mol each component in nitrogen
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
General description
Contents: 4 liters
Pressure: 120 psig
Outlet fitting: Aerosol-type push buttom
Weight: ~100 grams
Dimensions: 2.5 x 8 inches
D.O.T. specifications: 2Q
Application
Other Notes
Analyte
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1A - STOT RE 2 Inhalation
Storage Class Code
2B - Aerosol cans and lighters
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
501670:4548173983820
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service