Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

SML1825

Sigma-Aldrich

GSK984

≥98% (HPLC)

Synonym(s):

N-[(1S)-6-Chloro-2,3,4,9-tetrahydro-1H-carbazol-1-yl]-2-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16ClN3O
CAS Number:
Molecular Weight:
325.79
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -175 to -195°, c = 1 in methylene chloride

storage condition

protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(NC2=C3CCC[C@@H]2NC(C4=NC=CC=C4)=O)C3=C1

Biochem/physiol Actions

GSK984 is an inactive control probe for GSK983, which is an inhibitor of dihydroorotate dehydrogenase (DHODH) with antiviral activity. GSK983 blocks RNA virus replication with EC50 values of 10–40 nM, and also blocks cell proliferation. GSK984 acts by inhibition of dihydroorotate dehydrogenase (DHODH), necesssary for pyrimidine biosynthesis. Exogenous deoxycytidine was able to reduce GSK983 cytotoxicity, allowing cells to synthesize DNA, but still preventing RNA virus replication. GSK983 was active against dengue fever and Venezuelan equine encephalitis viruses. GSK984, the (S)-isomer, is almost 200-fold less active, so would make a good control.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

SML1825-5MG:
SML1825-25MG:
SML1825-VAR:
SML1825-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard M Deans et al.
Nature chemical biology, 12(5), 361-366 (2016-03-29)
Broad-spectrum antiviral drugs targeting host processes could potentially treat a wide range of viruses while reducing the likelihood of emergent resistance. Despite great promise as therapeutics, such drugs remain largely elusive. Here we used parallel genome-wide high-coverage short hairpin RNA
Robert Harvey et al.
Antiviral research, 82(1), 1-11 (2009-02-04)
GSK983, a novel tetrahydrocarbazole, inhibits the replication of a variety of unrelated viruses in vitro with EC(50) values of 5-20 nM. Both replication of the adenovirus Ad-5 and the polyoma virus SV-40, and episomal maintenance of human papillomaviruses (HPV) and
Kristjan S Gudmundsson et al.
Bioorganic & medicinal chemistry letters, 19(15), 4110-4114 (2009-06-27)
Synthesis of a series of tetrahydrocarbazole amides with potent activity against human papillomaviruses is described. Synthetic approaches allowing for variation of the substitution pattern of the tetrahydrocarbazole and the amide are outlined and resulting changes in antiviral activity and certain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service